Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Timm44
- Entrez Gene ID
-
21856
- Gene Name
- translocase of inner mitochondrial membrane 44
- Gene Aliases
-
0710005E20Rik, D8Ertd118e, Mimt44, Tim44
- Chromosome Location
-
Chr.8: 4259731 - 4275905 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC043479.1 |
- |
9 - 10 |
2315 |
103 |
View Full Product Details