Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Golim4
Entrez Gene ID
73124
Gene Name
golgi integral membrane protein 4
Gene Aliases
3110027H23Rik, GIMPC, GPP130, Golph4, P138
Chromosome Location
Chr.3: 75876183 - 75956949 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291069.1 NP_001277998.1 15 - 16 2553 124
NM_175193.6 NP_780402.1 14 - 15 2469 124
GenBank mRNA AK033730.1 - 14 - 15 2457 124
AK048434.1 - 15 - 16 2380 124
AK050346.1 - 2 - 3 137 124
AK078928.1 - 15 - 16 2553 124
BC023824.1 - 1 - 2 371 124
BC086642.1 - 14 - 15 2301 124
BC089354.1 - 14 - 15 2351 124

View Full Product Details