Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Spcs1
Entrez Gene ID
69019
Gene Name
signal peptidase complex subunit 1 homolog (S. cerevisiae)
Gene Aliases
1810004F21Rik
Chromosome Location
Chr.14: 30999826 - 31001666 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026911.3 NP_081187.2 2 - 3 524 92
GenBank mRNA AI848281.1 - 2 - 3 92
AK007336.1 - 2 - 3 222 92
AK082928.1 - 2 - 3 223 92
BC034270.2 - 2 - 3 399 93
BC049895.1 - 2 - 3 215 92

View Full Product Details