Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Spcs1
Entrez Gene ID
69019
Gene Name
signal peptidase complex subunit 1 homolog (S. cerevisiae)
Gene Aliases
1810004F21Rik
Chromosome Location
Chr.14: 30999826 - 31001666 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026911.3 NP_081187.2 3 - 4 611 67
GenBank mRNA AI848281.1 - 1 - 2 67
AK007336.1 - 3 - 4 309 67
AK082928.1 - 3 - 4 310 67
BC034270.2 - 3 - 4 486 67
BC049895.1 - 3 - 4 302 67

View Full Product Details