The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178730.3 | NP_848845.1 | 1 - 2 | 103 | 100 |
GenBank mRNA | AK028447.1 | - | 1 - 2 | 105 | 100 |
AK028587.1 | - | 1 - 2 | 97 | 101 | |
AK029032.1 | - | 1 - 2 | 65 | 100 | |
AK036957.1 | - | 1 - 2 | 104 | 100 |