The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033124.1 | NP_001028296.1 | 10 - 11 | 1279 | 76 |
GenBank mRNA | AK081588.1 | - | 10 - 11 | 1282 | 76 |
AK148691.1 | - | 3 - 4 | 188 | 76 |