Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rrnad1
Entrez Gene ID
229503
Gene Name
ribosomal RNA adenine dimethylase domain containing 1
Gene Aliases
-
Chromosome Location
Chr.3: 87922601 - 87930588 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153562.4 NP_705790.3 2 - 3 463 104
GenBank mRNA AK035389.1 - 2 - 3 415 104
AK082223.1 - 2 - 3 396 104

View Full Product Details