Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc10a4
Entrez Gene ID
231290
Gene Name
solute carrier family 10 (sodium/bile acid cotransporter family), member 4
Gene Aliases
E130304D01
Chromosome Location
Chr.5: 73006904 - 73012955 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173403.2 NP_775579.2 1 - 2 752 59
GenBank mRNA AK087479.1 - 1 - 2 753 59
AK149202.1 - - 753 59
BC132529.1 - 1 - 2 664 59
BC138048.1 - 1 - 2 664 59

View Full Product Details