Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sgcb
Entrez Gene ID
24051
Gene Name
sarcoglycan, beta (dystrophin-associated glycoprotein)
Gene Aliases
43kDa, AI747103, AI844814, beta-SG
Chromosome Location
Chr.5: 73632749 - 73647731 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011890.4 NP_036020.1 1 - 2 59 91
GenBank mRNA AB024921.1 - 1 - 2 73 91
AK014381.2 - 1 - 2 73 91
AK132306.1 - 1 - 2 118 91
AK169403.1 - 1 - 2 60 91

View Full Product Details