Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Spata18
Entrez Gene ID
73472
Gene Name
spermatogenesis associated 18
Gene Aliases
1700067I02Rik
Chromosome Location
Chr.5: 73651380 - 73679484 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178387.3 NP_848474.2 11 - 12 1764 73
GenBank mRNA AK006916.1 - 10 - 11 1443 73
BC120503.1 - 8 - 9 1216 73
BC120505.1 - 8 - 9 1216 73
BC144721.1 - 8 - 9 1216 73

View Full Product Details