Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
Sh3bp5
Entrez Gene ID
24056
Gene Name
SH3-domain binding protein 5 (BTK-associated)
Gene Aliases
AI606498, SH3BP-5, Sab
Chromosome Location
Chr.14: 31373774 - 31436033 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK043375.1 - - 1590 82

View Full Product Details