The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001284346.1 | NP_001271275.1 | 2 - 3 | 355 | 72 |
NM_172254.4 | NP_758458.1 | 2 - 3 | 355 | 72 | |
NR_104303.1 | 2 - 3 | 213 | 72 | ||
NR_104304.1 | 2 - 3 | 481 | 72 | ||
NR_104305.1 | 1 - 2 | 377 | 72 | ||
GenBank mRNA | AK011448.1 | - | 2 - 3 | 273 | 72 |
AK017764.1 | - | 2 - 3 | 399 | 72 | |
AK048097.1 | - | 2 - 3 | 245 | 72 | |
BC029910.1 | - | 2 - 3 | 246 | 72 | |
BC039954.1 | - | 2 - 3 | 266 | 72 | |
BG082470.2 | - | 1 - 2 | 294 | 72 |