Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pglyrp4
Entrez Gene ID
384997
Gene Name
peptidoglycan recognition protein 4
Gene Aliases
Gm1478, Gm413
Chromosome Location
Chr.3: 90726064 - 90741517 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001165968.1 NP_001159440.1 7 - 8 992 99
NM_207263.2 NP_997146.2 7 - 8 989 99
GenBank mRNA AK137756.1 - 2 - 3 255 99
AY518699.1 - 7 - 8 943 99
BC120840.1 - 7 - 8 984 99
BC144902.1 - 7 - 8 992 99
BC144927.1 - 6 - 7 873 99

View Full Product Details