Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ncapg
Entrez Gene ID
54392
Gene Name
non-SMC condensin I complex, subunit G
Gene Aliases
5730507H05Rik, Hcapg, MFT.M05.13
Chromosome Location
Chr.5: 45669873 - 45700547 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019438.1 NP_062311.1 16 - 17 2552 143
GenBank mRNA AJ237585.1 - 10 - 11 2946 132
AK136254.1 - 11 - 12 1538 143
AK138026.1 - 1 - 2 1871 143
BC137834.1 - 9 - 10 1770 143
BC144960.1 - 9 - 10 1770 143

View Full Product Details