Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lyar
Entrez Gene ID
17089
Gene Name
Ly1 antibody reactive clone
Gene Aliases
MLZ-264
Chromosome Location
Chr.5: 38220482 - 38234306 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025281.3 NP_079557.2 2 - 3 323 94
GenBank mRNA AK006373.1 - 2 - 3 257 94
AK076727.1 - 2 - 3 328 94
AK211108.1 - 3 - 4 279 94
BC116923.1 - 1 - 2 179 94
BC116927.1 - 1 - 2 179 94

View Full Product Details