Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zbtb49
Entrez Gene ID
75079
Gene Name
zinc finger and BTB domain containing 49
Gene Aliases
4930518A03Rik, Zfp509, Znf509
Chromosome Location
Chr.5: 38189732 - 38220449 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029162.2 NP_083438.1 4 - 5 1391 97
GenBank mRNA AK015819.1 - 2 - 3 303 98
AK043044.1 - 4 - 5 1386 97
AK140546.1 - 4 - 5 1386 97


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK148068.1 - 1397 Chr. - - [-] on Build 97

View Full Product Details