Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mx2
Entrez Gene ID
17858
Gene Name
MX dynamin-like GTPase 2
Gene Aliases
AI528743, Mx-2
Chromosome Location
Chr.16: 97536081 - 97560901 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_003508.1 8 - 9 1108 82
GenBank mRNA AK143831.1 - 8 - 9 1109 82
AK147949.1 - - 1728 82
BC007127.1 - 8 - 9 82
J03368.1 - 8 - 9 1106 82


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AA183899.1 - 3 - 4 321 Chr. 16 - 97546843 - 97552068 [+] on Build GRCm38 82
GenBankmRNA BI149262.1 - 2 - 3 Chr. 16 - 97547457 - 97558673 [+] on Build GRCm38 82
GenBankmRNA BF143741.1 - Chr. - - [-] on Build 80
GenBankmRNA CA544052.1 - 2 - 3 479 Chr. 16 - 97550387 - 97551970 [+] on Build GRCm38 82


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream family with sequence similarity 3, member B Fam3b 2-21, 9030624C24Rik, D16Jhu19e, ORF9, Pander -46.0kb Anti-sense Chr. - 97471086 - 97504936 on Build GRCm38
Downstream transmembrane protease, serine 2 Tmprss2 D16Ertd61e 13.2kb Anti-sense Chr. - 97564682 - 97659538 on Build GRCm38

View Full Product Details