Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ptpn20
Entrez Gene ID
19256
Gene Name
protein tyrosine phosphatase, non-receptor type 20
Gene Aliases
typ
Chromosome Location
Chr.14: 33589201 - 33640754 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008978.2 NP_033004.1 6 - 7 582 84
GenBank mRNA AK017003.1 - 6 - 7 645 84
AK029493.1 - 6 - 7 580 84
D64141.1 - 5 - 6 581 84

View Full Product Details