The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290383.1 | NP_001277312.1 | 10 - 11 | 1578 | 97 |
NM_194055.3 | NP_918944.2 | 10 - 11 | 1581 | 97 | |
GenBank mRNA | AK042164.1 | - | 10 - 11 | 1193 | 97 |
AK140848.1 | - | 10 - 11 | 1579 | 97 | |
BC031468.1 | - | 8 - 9 | 884 | 97 | |
BC059280.1 | - | 8 - 9 | 882 | 97 | |
DV654302.1 | - | 5 - 6 | 642 | 97 |