Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Schip1
Entrez Gene ID
30953
Gene Name
schwannomin interacting protein 1
Gene Aliases
Nf2ip, Schip-1
Chromosome Location
Chr.3: 68064802 - 68626482 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC094348.1 - 1 - 2 112 77

View Full Product Details