Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Birc7
Entrez Gene ID
329581
Gene Name
baculoviral IAP repeat-containing 7 (livin)
Gene Aliases
E130019N06, KIAP, Livin, ML-IAP
Chromosome Location
Chr.2: 180928152 - 180935028 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163247.1 NP_001156719.1 2 - 3 590 68
GenBank mRNA BC107260.2 - 2 - 3 514 68
BC125013.1 - 2 - 3 514 68

View Full Product Details