The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163247.1 | NP_001156719.1 | 4 - 5 | 717 | 85 |
GenBank mRNA | AK053463.1 | - | 3 - 4 | 1071 | 85 |
BC107260.2 | - | 4 - 5 | 641 | 85 | |
BC125013.1 | - | 4 - 5 | 641 | 85 |