The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020560.2 | NP_065585.1 | 1 - 2 | 243 | 98 |
GenBank mRNA | AK005110.1 | - | 1 - 2 | 202 | 98 |
AK011825.1 | - | 1 - 2 | 199 | 98 | |
AK019202.1 | - | 1 - 2 | 223 | 98 | |
AK078073.1 | - | 1 - 2 | 186 | 98 | |
AK161913.1 | - | 1 - 2 | 186 | 98 | |
BC006764.1 | - | 1 - 2 | 138 | 98 | |
BC027430.1 | - | 1 - 2 | 147 | 98 | |
BY237441.1 | - | 1 - 2 | 245 | 98 | |
Z46966.1 | - | 1 - 2 | 162 | 98 |