Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Clock
Entrez Gene ID
12753
Gene Name
circadian locomotor output cycles kaput
Gene Aliases
5330400M04Rik, KAT13D
Chromosome Location
Chr.5: 76209868 - 76305007 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289826.1 NP_001276755.1 19 - 20 2032 61
NM_007715.6 NP_031741.1 19 - 20 2075 61
GenBank mRNA AB221635.1 - 16 - 17 1687 61
AF000998.1 - 19 - 20 2075 61
AK129118.1 - 19 - 20 2032 61
AY118227.1 - 4 - 5 437 61
BC129886.1 - 17 - 18 1783 61

View Full Product Details