Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pdcl2
Entrez Gene ID
79455
Gene Name
phosducin-like 2
Gene Aliases
1700010B22Rik, 1700016K07Rik, Mgcphlp, Tphlp
Chromosome Location
Chr.5: 76312127 - 76331172 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023508.6 NP_075997.1 2 - 3 219 64
GenBank mRNA AK005602.1 - 2 - 3 242 64
AK005814.1 - 2 - 3 219 64
AK006773.1 - 2 - 3 236 64
AK132738.1 - 2 - 3 266 64
BC048432.1 - 2 - 3 221 64

View Full Product Details