Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rgr
Entrez Gene ID
57811
Gene Name
retinal G protein coupled receptor
Gene Aliases
-
Chromosome Location
Chr.14: 37034909 - 37049014 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021340.4 NP_067315.1 2 - 3 317 59
GenBank mRNA AF076930.1 - 2 - 3 237 59
AK044450.1 - 2 - 3 269 59
AK044649.1 - 2 - 3 268 59
AK149272.1 - 2 - 3 268 59
BC082611.1 - 2 - 3 254 59
BC086689.1 - 2 - 3 244 59
BC119055.1 - 2 - 3 251 59
CK622127.1 - 2 - 3 317 59

View Full Product Details