Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fcrl5
Entrez Gene ID
329693
Gene Name
Fc receptor-like 5
Gene Aliases
Fcrh3, mBXMH2
Chromosome Location
Chr.3: 87435782 - 87500678 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_183222.3 NP_899045.3 2 - 3 193 74
GenBank mRNA AY158090.1 - 2 - 3 193 74
AY506558.1 - 2 - 3 202 74
BC137997.1 - 2 - 3 130 74
BC137998.1 - 2 - 3 130 74

View Full Product Details