Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pcdh11x
Entrez Gene ID
245578
Gene Name
protocadherin 11 X-linked
Gene Aliases
PCDHX, PCDHX11, Pcdh11
Chromosome Location
Chr.X: 120290264 - 120910622 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271809.1 NP_001258738.1 7 - 7 4322 77
NM_001271810.1 NP_001258739.1 6 - 6 4292 77
NR_073451.1 8 - 8 4468 77
NR_073452.1 7 - 7 4438 77
GenBank mRNA AY861426.1 - 6 - 6 1597 77
AY861427.1 - 5 - 5 1567 77
AY861428.1 - 5 - 5 1451 77
AY861429.1 - 4 - 4 1421 77
BC126930.1 - 7 - 7 4361 77

View Full Product Details