The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027322.2 | NP_081598.1 | 1 - 2 | 131 | 69 |
GenBank mRNA | AK008151.1 | - | 1 - 2 | 89 | 69 |
AK161802.1 | - | 1 - 2 | 130 | 69 | |
CJ052035.1 | - | 1 - 2 | 129 | 69 | |
CX242928.1 | - | 1 - 2 | 82 | 69 |