Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ythdc1
Entrez Gene ID
231386
Gene Name
YTH domain containing 1
Gene Aliases
A730098D12Rik, C80342, mKIAA1966
Chromosome Location
Chr.5: 86803980 - 86836659 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177680.3 NP_808348.2 14 - 15 1862 74
GenBank mRNA AB221619.1 - 6 - 7 462 74
AK080576.1 - 1 - 2 74 74
AK173312.1 - 10 - 11 3533 74
BC022697.1 - 7 - 8 631 74
BC037593.1 - 8 - 9 2089 74
BC044650.1 - 10 - 11 5139 74
BC048817.1 - 7 - 8 616 74
DV042054.1 - 2 - 3 153 74

View Full Product Details