Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Best1
Entrez Gene ID
24115
Gene Name
bestrophin 1
Gene Aliases
Bmd, Vmd2, mBest1
Chromosome Location
Chr.19: 9985008 - 10001653 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011913.2 NP_036043.2 3 - 4 458 92
GenBank mRNA AK006549.1 - 1 - 2 78 92
AY450427.1 - 2 - 3 282 92
BC139060.1 - 2 - 3 273 92
BC139061.1 - 2 - 3 273 92
BY714344.1 - 3 - 4 432 92

View Full Product Details