Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sh3bgr
Entrez Gene ID
50795
Gene Name
SH3-binding domain glutamic acid-rich protein
Gene Aliases
5430437A18Rik
Chromosome Location
Chr.16: 96200470 - 96228937 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015825.2 NP_056640.1 4 - 5 617 89
GenBank mRNA AJ239082.1 - 4 - 5 383 89
AJ272170.1 - 4 - 5 445 89
BC055870.1 - 4 - 5 434 89

View Full Product Details