The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_199467.2 | NP_955761.2 | 10 - 11 | 1623 | 153 |
GenBank mRNA | AK089531.1 | - | 3 - 4 | 363 | 153 |
AK166072.1 | - | 10 - 11 | 1634 | 153 | |
AY512923.1 | - | 5 - 6 | 757 | 153 | |
BC094429.1 | - | 9 - 10 | 967 | 153 | |
BC137910.1 | - | 5 - 6 | 533 | 153 | |
BC145028.1 | - | 5 - 6 | 533 | 153 |