Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Aco1
Entrez Gene ID
11428
Gene Name
aconitase 1
Gene Aliases
Aco-1, Irebp, Irp1
Chromosome Location
Chr.4: 40143265 - 40199009 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007386.2 NP_031412.2 4 - 5 710 86
GenBank mRNA AK027914.1 - 4 - 5 496 86
AK085951.1 - 4 - 5 459 86
AK129025.1 - 4 - 5 492 86
AK163985.1 - 4 - 5 509 86
BC005454.1 - 4 - 5 470 86
X61147.1 - 4 - 5 477 86

View Full Product Details