Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pls1
Entrez Gene ID
102502
Gene Name
plastin 1 (I-isoform)
Gene Aliases
AI427122
Chromosome Location
Chr.9: 95752642 - 95845306 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033210.3 NP_001028382.1 13 - 14 1653 144
GenBank mRNA AK030875.1 - 6 - 7 651 144
AK133070.1 - 13 - 14 1635 144
BC026410.1 - 3 - 4 328 144
BC139068.1 - 12 - 13 1555 144
BC139069.1 - 12 - 13 1555 144

View Full Product Details