The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033210.3 | NP_001028382.1 | 7 - 8 | 887 | 118 |
GenBank mRNA | AK133070.1 | - | 7 - 8 | 869 | 118 |
BC139068.1 | - | 6 - 7 | 789 | 118 | |
BC139069.1 | - | 6 - 7 | 789 | 118 |