The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_199018.2 | NP_950183.1 | 10 - 11 | 2702 | 80 |
GenBank mRNA | AK133927.1 | - | 10 - 11 | 2650 | 80 |
AK149830.1 | - | 10 - 11 | 2633 | 80 | |
AK150259.1 | - | 10 - 11 | 2633 | 80 | |
AK172910.1 | - | 1 - 2 | 124 | 80 | |
BC023434.1 | - | 2 - 3 | 226 | 80 | |
BC034186.1 | - | 6 - 7 | 1824 | 80 | |
BC062944.1 | - | 10 - 11 | 2617 | 80 | |
BC078461.1 | - | 9 - 10 | 2465 | 80 |