The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015794.1 | NP_056609.1 | 2 - 3 | 1673 | 133 |
GenBank mRNA | AF176529.1 | - | 2 - 3 | 393 | 133 |
AK154497.1 | - | 2 - 3 | 182 | 133 |