Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmcc3
Entrez Gene ID
319880
Gene Name
transmembrane and coiled coil domains 3
Gene Aliases
AW488095, C630016B22Rik, C88213, Tmcc1
Chromosome Location
Chr.10: 94311912 - 94590954 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172051.3 NP_742048.2 1 - 2 246 85
GenBank mRNA AK083129.1 - 1 - 2 149 85
AK132545.1 - 1 - 2 224 85
BB619673.1 - 1 - 2 247 85
BC095951.1 - 1 - 2 180 85

View Full Product Details