Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pibf1
Entrez Gene ID
52023
Gene Name
progesterone immunomodulatory binding factor 1
Gene Aliases
-
Chromosome Location
Chr.14: 99099424 - 99254495 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029320.3 NP_083596.2 17 - 18 2938 109
GenBank mRNA AK006060.1 - 8 - 9 912 109
BC038269.1 - 4 - 5 544 108

View Full Product Details