The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028721.3 | NP_082997.3 | 12 - 13 | 2113 | 72 |
GenBank mRNA | AK133217.1 | - | 12 - 13 | 2114 | 72 |
AK173327.1 | - | 12 - 13 | 1692 | 72 | |
AY259499.1 | - | 12 - 13 | 2053 | 72 | |
BC115724.1 | - | 12 - 13 | 1643 | 72 |