The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033314.3 | NP_201571.2 | 4 - 5 | 746 | 66 |
GenBank mRNA | AF323958.1 | - | 4 - 5 | 615 | 66 |
AK166724.1 | - | 4 - 5 | 747 | 66 | |
AK169706.1 | - | 4 - 5 | 749 | 66 | |
BC035200.1 | - | - | 736 | 66 |