The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177778.2 | NP_001171249.1 | 18 - 19 | 2660 | 103 |
NM_001177779.1 | NP_001171250.1 | 12 - 13 | 1657 | 103 | |
NM_001177780.1 | NP_001171251.1 | 10 - 11 | 1049 | 103 | |
NM_001290402.1 | NP_001277331.1 | 17 - 18 | 2606 | 103 | |
NM_016747.4 | NP_058027.1 | 19 - 20 | 2702 | 103 | |
GenBank mRNA | AK122474.1 | - | 18 - 19 | 2659 | 103 |
AK145815.1 | - | 12 - 13 | 1412 | 103 | |
AK148464.1 | - | 10 - 11 | 1029 | 103 | |
AK159655.1 | - | 10 - 11 | 1037 | 103 | |
AK167365.1 | - | 10 - 11 | 1036 | 103 | |
BC060713.1 | - | 17 - 18 | 2734 | 103 | |
BC094368.1 | - | 17 - 18 | 2601 | 103 | |
D87117.1 | - | 19 - 20 | 2676 | 103 |