The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146690.1 | NP_001140162.1 | 6 - 7 | 1052 | 95 |
NM_144807.3 | NP_659056.3 | 6 - 7 | 1052 | 95 | |
NR_027477.1 | 6 - 7 | 1052 | 95 | ||
GenBank mRNA | AK032497.1 | - | 6 - 7 | 947 | 95 |
AY445814.1 | - | 6 - 7 | 947 | 95 | |
BE650891.1 | - | 2 - 3 | 287 | 95 |