The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286647.1 | NP_001273576.1 | 5 - 6 | 557 | 103 |
NM_019717.3 | NP_062691.3 | 6 - 7 | 802 | 103 | |
NM_178050.4 | NP_835151.2 | 5 - 6 | 557 | 103 | |
GenBank mRNA | AK038697.1 | - | 5 - 6 | 485 | 103 |
AK135988.1 | - | 5 - 6 | 448 | 103 | |
BC055466.1 | - | 3 - 4 | 160 | 103 | |
BC060501.1 | - | 6 - 7 | 707 | 103 |