Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tsx
Entrez Gene ID
22127
Gene Name
testis specific X-linked gene
Gene Aliases
AV207184
Chromosome Location
Chr.X: 103409101 - 103424588 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009440.2 NP_033466.1 2 - 3 219 78
GenBank mRNA BC048493.1 - 2 - 3 219 78
HQ433540.1 - 3 - 4 330 78
HQ433541.1 - 3 - 4 434 78
HQ433542.1 - 3 - 4 329 78
HQ433544.1 - 2 - 3 519 78
HQ433547.1 - 2 - 3 419 78
X99796.1 - 2 - 3 204 78

View Full Product Details