Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kcnq5
Entrez Gene ID
226922
Gene Name
potassium voltage-gated channel, subfamily Q, member 5
Gene Aliases
7730402H11, 9230107O05Rik, AA589396, D1Mgi1
Chromosome Location
Chr.1: 21398399 - 21962650 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023872.3 NP_076361.1 9 - 10 1613 70
GenBank mRNA AK033079.1 - 3 - 4 171 70
AY679158.1 - 9 - 10 1252 70

View Full Product Details