The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008929.3 | NP_032955.2 | 1 - 2 | 243 | 60 |
GenBank mRNA | AK146097.1 | - | 1 - 2 | 207 | 60 |
AK148270.1 | - | 1 - 2 | 200 | 60 | |
AK212342.1 | - | 1 - 2 | 59 | 60 | |
BB618718.1 | - | 1 - 2 | 243 | 60 | |
BC013766.1 | - | 1 - 2 | 188 | 60 | |
U28423.1 | - | 1 - 2 | 186 | 60 |