Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
E2f7
Entrez Gene ID
52679
Gene Name
E2F transcription factor 7
Gene Aliases
A630014C11Rik, D10Ertd739e
Chromosome Location
Chr.10: 110745427 - 110787384 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178609.4 NP_848724.2 2 - 3 304 68
GenBank mRNA AK031078.1 - 2 - 3 302 68
AK041481.1 - 2 - 3 302 68
AK045040.1 - 2 - 3 269 68
AK051521.1 - 2 - 3 304 68
AK143686.1 - 2 - 3 277 68
AY463359.1 - 2 - 3 301 68
BC145429.1 - 2 - 3 257 68
BC150772.1 - 2 - 3 257 68

View Full Product Details