Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zdhhc17
Entrez Gene ID
320150
Gene Name
zinc finger, DHHC domain containing 17
Gene Aliases
A230053P19Rik, BB187739, D130071N24Rik, Hip14
Chromosome Location
Chr.10: 110941779 - 111010066 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172554.2 NP_766142.2 11 - 12 1305 153
GenBank mRNA AK039575.1 - 11 - 12 1306 153
AK046891.1 - 11 - 12 1247 153
BC027101.1 - 11 - 12 1193 153
BC117788.1 - 11 - 12 1247 153
BC117789.2 - 11 - 12 1247 153

View Full Product Details